EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Two diacylated malvidin glycosides from Petunia hybrida flowers.
PMID 11738418
Abstract
Two novel diacylated and two known anthocyanins were isolated from violet flowers of Petunia hybrida cv Festival. The new anthocyanins are malvidin 3-O-(6-O-(4-O-(4-O-(6-O-feruloyl-beta-D-glucopyranosyl)-E-p-coumaroyl)-alpha-rhamnosyl)-beta-D-glucopyranoside)-5-beta-D-glucopyranoside and malvidin 3-O-(6-O-(4-O-(4-O-(6-O-E-p-coumaroyl-beta-D-glucopyranosyl)-E-p-coumaroyl)-alpha-rhamnosyl)-beta-D-glucopyranoside)-5-beta-D-glucopyranoside. The two known pigments are the 3-caffeoylglucosyl-p-coumaroylrutinoside-5-glucosides of malvidin and petunidin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68120
Malvidin chloride, ≥95.0% (HPLC)
C17H15ClO7