EMAIL THIS PAGE TO A FRIEND
Journal of HIV therapy
From amprenavir to GW433908.
PMID 11740410
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0685 Amprenavir, ≥98% (HPLC)
C25H35N3O6S