EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A synthetic membrane-anchored antigen efficiently promotes uptake of antifluorescein antibodies and associated protein a by mammalian cells.
PMID 11741450
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201626
Fluoresceinamine, isomer I
C20H13NO5