EMAIL THIS PAGE TO A FRIEND
Journal of applied toxicology : JAT
In vitro and in vivo genetic toxicology studies with diethylene glycol monohexyl ether.
PMID 11746191
Abstract
Diethylene glycol monohexyl ether (DEGHE; CAS no. 112-59-4), an industrial chemical, was investigated for the potential to produce genotoxic effects using three in vitro and two in vivo tests. No mutagenic activity occurred in either the absence or presence of metabolic activation with a Salmonella typhimurium reverse assay using strains TA98, TA100, TA1535, TA1537 and TA1538. In a Chinese hamster ovary (CHO) forward gene mutation test (HGPRT locus) there was an increase in the mutation frequencies, which were relatively small compared with the solvent control values, somewhat inconsistent between duplicate cultures and occurred particularly in the presence of metabolic activation. Linear regression analysis indicated a marginally significant trend for dosage versus mutation frequency, suggesting that DEGHE was weakly positive in this test. A sister chromatid exchange test in CHO cells showed no significant dosage-related effects in the presence or absence of metabolic activation. A peripheral blood micronucleus test in mice by dosing with an intraperitoneal injection of DEGHE did not show any potential for DEGHE to increase the incidence of micronucleated polychromatophilic erythrocytes. In a first femoral bone marrow chromosome aberration test in the rat by peroral dosing, DEGHE did not cause any increase in aberrations for 12-h and 24-h samples with males and females or with females at 48-h sampling. However, with males at 48 h the two lowest doses showed an increased number of aberrations, but not at the high doses. A repeat study in males with a larger number of doses and 24-h and 48-h samples did not replicate this finding. It is concluded that DEGHE may have limited weak mutagenic activity in vitro but is devoid of clastogenic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
449393
Di(ethylene glycol) hexyl ether, 95%
C10H22O3