EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Direct detection of Ziram by liquid chromatography/atmospheric pressure chemical ionization mass spectrometry.
PMID 11746904
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329711
Zinc dimethyldithiocarbamate, 97%
C6H12N2S4Zn
45708
Ziram, PESTANAL®, analytical standard
C6H12N2S4Zn