EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Anti-MRSA activity of alkyl gallates.
PMID 11755333
Abstract
A series of alkyl gallates (3,4,5-trihydroxybenzoates) was found to show antibacterial activity against Gram-positive bacteria including methicillin resistant Staphylococcus aureus (MRSA) strains. For example, dodecyl (C(12)) gallate (1) exhibited bactericidal activity against MRSA ATCC 33591 strain with the minimum bactericidal concentration (MBC) of 25 microg/mL (74 microM). The time-kill curve study showed that dodecyl gallate is bactericidal against this MRSA strain. This bactericidal activity comes in part from its ability to inhibit respiratory electron transport systems. The length of the alkyl chain is not a major contributor but plays an important role in eliciting the activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48660
Lauryl gallate, antioxidant, ≥99.0% (HPLC)
C19H30O5
53024
Lauryl gallate, analytical standard
C19H30O5