EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and structure-activity relationship of 2-(aminoalkyl)-2,3,3a,8-tetrahydrodibenzo[c,f]isoxazolo[2,3-a]azepine derivatives: a novel series of 5-HT(2A/2C) receptor antagonists. Part 1.
PMID 11755364
Abstract
The synthesis of a series of novel 2-(aminoalkyl)-2,3,3a,8-tetrahydrodibenzo[c,f]isoxazolo[2,3-a]azepine derivatives as well as their 5-HT(2A/2C) and H(1) receptor binding affinities are described. The in vivo activity as potential anxiolytics of the synthesised compounds was measured in a mCPP challenge test. One of the compounds, 2a, proved to be a potent 5-HT(2A/2C) receptor antagonist showing as well oral activity and therefore could be considered as a potential anxiolytic/antidepressant agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235350
2-Benzylaniline, ≥99%
C13H13N