EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
Fluorescent labeling of drugs and simple organic compounds containing amine functional groups, utilizing dansyl chloride in Na(2)CO(3) buffer.
PMID 11755390
Abstract
Fluorescent labeling of amine functional groups using dansyl chloride (DNS-Cl), and in sodium carbonate buffer, allowed the detection of 1 microg amounts of analytes. The methodology presented allows dansylation of primary, secondary, and tertiary amine groups at a temperature of 25 degrees C. The dansylation of tertiary amines involves a chemical reaction which removes one substituent (or branch) of the amine group. A one molar working concentration of Na(2)CO(3) is used, and is at pH 11.0. Compounds such as isopropylamine, dipropylamine, diethylamine, triethylamine, triisooctylamine, and N,N-dimethylaniline were labeled by use of DNS-Cl from samples obtained from a complex mixture of alkanes. The compound p-chloroaniline contains a primary amine group and is a solid at 25 degrees, quickly dissolves in the one molar sodium carbonate buffer and is dansylated in 15 min. Heroin, which contains a tertiary amine group, was extracted into ethyl acetate from an aqueous solution, then reacted with DNS-Cl. Benzocaine, a local anesthetic, was dansylated in 15 min. Tertiary amine groups incorporated in a rigid ring system, such as for caffeine, strychnine, and the ionic salt form of cocaine hydrochloride did not react with DNS-Cl under these conditions. The reaction time for tertiary amines was 2 h or less, and 15 min for compounds having primary and secondary amine groups. Separation of the dansylated compounds from unreacted DNS-Cl was accomplished by diethyl ether extraction of the aqueous reaction solution, followed by thin layer chromatography using various organic solvents such as acetone and methylene chloride.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T81000
Trioctylamine, 98%
C24H51N
92830
Trioctylamine, purum, ≥98.0% (GC)
C24H51N