EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Co-metabolic degradation of chlorinated hydrocarbons by Pseudomonas sp. strain DCA1.
PMID 11762603
Abstract
Pseudomonas sp. strain DCA1, which is capable of utilizing 1,2-dichloroethane (DCA) as sole carbon and energy source, was used to oxidize chlorinated methanes, ethanes, propanes, and ethenes. Chloroacetic acid, an intermediate in the DCA degradation pathway of strain DCA1, was used as a co-substrate since it was readily oxidized by DCA-grown cells of strain DCAI and did not compete for the monooxygenase. All of the tested compounds except tetrachloroethylene (PER) were oxidized by cells expressing DCA monooxygenase. Strain DCAI could not utilize any of these compounds as a growth substrate. Co-metabolic oxidation during growth on DCA was studied with 1,2-dichloropropane. Although growth on this mixture occurred, 1,2-dichloropropane strongly inhibited growth of strain DCAI. This inhibition was not caused by competition for the monooxygenase. It was shown that the oxidation of 1,2dichloropropane resulted in the accumulation of 2,3-dichloro-1-propanol and 2-chloroethanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02577
1,2-Dichloropropane, analytical standard
C3H6Cl2
82270
1,2-Dichloropropane, 99%
C3H6Cl2