EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Identification of hepatic protein targets of the reactive metabolites of the non-hepatotoxic regioisomer of acetaminophen, 3'-hydroxyacetanilide, in the mouse in vivo using two-dimensional gel electrophoresis and mass spectrometry.
PMID 11765013
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7205
3-Acetamidophenol, 97%
C8H9NO2