EMAIL THIS PAGE TO A FRIEND
Mycoses
Otomycosis in Nigeria: treatment with mercurochrome.
PMID 11766105
Abstract
Seventy-two mycologically proven cases of otomycosis (38 males, 34 females) aged 8-80 years were investigated. Aspergillus niger was the commonest aetiological agent (43.1%) followed by Candida spp. (22.2%), A. flavus (19.4%) and A. fumigatus (15.3%). Three topical drugs, namely mercurochrome, clotrimazole and locacorten-vioform, were evaluated for otomycosis therapy in separate groups of 24 patients each. Mercurochrome was found to be the most efficacious in terms of healing, relief from symptoms of the disease and production of negative fungal cultures. Mercurochrome is recommended as a safe and economical drug for the topical treatment of otomycosis in developing countries like Nigeria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7011
Mercury dibromofluorescein disodium salt, practical grade, crystalline
C20H8Br2HgNa2O6