EMAIL THIS PAGE TO A FRIEND
Cutis
Botanical briefs: The cashew tree--Anacardium occidentale L.
PMID 11766115
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91822
5-Pentadecylresorcinol, analytical standard
C21H36O2