EMAIL THIS PAGE TO A FRIEND
Research in microbiology
Characterization of recombinant Bordetella pertussis adenylate cyclase toxins carrying passenger proteins.
PMID 11766964
Abstract
Bordetella pertussis secretes a calmodulin-activated adenylate cyclase toxin, CyaA, that is able to deliver its N-terminal catalytic domain (400 amino acid residues) into the cytosol of eukaryotic target cells, directly through the cytoplasmic membrane. We have previously shown that CyaA can be used as a vehicle to deliver CD8+ T-cell epitopes, inserted within the catalytic domain of the toxin, into antigen-presenting cells and can trigger specific class I-restricted cytotoxic T-cell (CTL) responses in vivo. To explore the tolerance of CyaA to insertion of polypeptides of larger size, we constructed and characterized different recombinant CyaA toxins with protein inserts of 87 to 206 amino acids in length. Several of these recombinant CyaA toxins were found to be invasive. Furthermore, we showed that the unfolding of the passenger protein is a prerequisite for the translocation of the recombinant toxins into eukaryotic cells. Our results highlight the remarkable tolerance of the CyaA toxin and suggest that CyaA might be used to deliver proteins into eukaryotic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0389 Restrictocin from Aspergillus restrictus, lyophilized powder