EMAIL THIS PAGE TO A FRIEND
Environmental and molecular mutagenesis
Point mutations induced by 1,2-epoxy-3-butene N1 deoxyinosine adducts.
PMID 11774359
Abstract
The National Toxicology Program has recently classified 1,3-butadiene (BD) as a human carcinogen. BD is metabolized to the intermediates 1,2-epoxy-3-butene (EB), 1,2:3,4-diepoxybutane (DEB), and 1,2-dihydroxy-3,4-epoxybutane. All three metabolites have been implicated in producing specific types of DNA damage and as genotoxic agents in mice, rat, and human cells. This study has focused on EB-induced N1 deoxyinosine lesions that are formed by deamination of deoxyadenosine following reaction of the epoxide at the N(1) position. The R and S stereoisomers of this lesion were incorporated site-specifically within the context of an 11-mer oligodeoxynucleotide, incorporated into M13mp7L2 single-stranded DNA, and transfected into E. coli. Both stereoisomers modestly reduced plaque-forming ability, indicating that neither lesion presents a base modification that cannot be bypassed. The resulting plaques were assessed for point mutations using differential hybridization and DNA sequence analyses. The overall mutagenic spectrum revealed that the N1 adducts were highly mutagenic (approximately 90% per replication cycle), causing a predominance of A --> G transitions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
127574
3,4-Epoxy-1-butene, 98%
C4H6O