EMAIL THIS PAGE TO A FRIEND
Journal of the Egyptian Society of Parasitology
Efficacy of flumethrin and coumaphos against the camel tick Hyalomma dromedarii L. (Acari: Ixodidae).
PMID 11775105
Abstract
The efficacy of Flumethrin pour on and Coumaphos 50% WP was tested against different stages of Hyalomma dromedarii. With contact method, LC50 values for larvae and adults were 0.04, 0.03 ug/cm2 and 0.05, 1.06 ug/cm2 respectively. The LC50 for coumaphos against the larvae, adults and eggs using dipping method were 44, 63, and 62 ppm respectively. Flumethrin caused significant reduction in oviposition and hatchability in female ticks that survived sublethal concentrations, while sublethal doses of coumaphos had no effect on the reproductive potential of the tick. The results showed that flumethrin was 8 times more toxic than coumaphos.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46417
Flumethrin, PESTANAL®, analytical standard
C28H22Cl2FNO3