EMAIL THIS PAGE TO A FRIEND
Cancer research
Quantitation of metabolites of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone after cessation of smokeless tobacco use.
PMID 11782369
Abstract
Two major metabolites of the tobacco-specific lung carcinogen 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) were previously shown to be highly persistent in human urine after cessation of cigarette smoking. We hypothesized that NNK or its metabolite, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL), was sequestered in the lung. In this study, we further evaluated this hypothesis by quantifying the NNK metabolites, NNAL and its glucuronides (NNAL-Gluc), in urine and plasma after cessation of smokeless tobacco use, in which NNK is administered p.o. rather than by inhalation. Thirteen male nonsmokers, 11 snuff dippers and 2 tobacco chewers, participated in the study. Urine and plasma were obtained at baseline and at intervals 2-126 days after cessation of smokeless tobacco use. The distribution half-lives t(1/2alpha) (days) of NNAL (1.32 +/- 0.85 versus 3.35 +/- 1.86) and NNAL-Gluc (1.53 +/- 1.22 versus 3.89 +/- 2.43) were significantly shorter in smokeless tobacco users than in smokers. There were no significant differences in the terminal half-lives t(1/2beta) (days) of NNAL (26.3 +/- 16.7 versus 45.2 +/- 26.9) and NNAL-Gluc (26.1 +/- 15.1 versus 39.6 +/- 26.0) in smokeless tobacco users and smokers. Baseline levels as well as renal clearance of the NNK metabolites correlated with number of tins or pouches of smokeless tobacco consumed. Ratios of (S)-NNAL:(R)-NNAL and (S)-NNAL-Gluc:(R)-NNAL-Gluc in urine were significantly (3.1-5.7 times) higher 7 days after cessation than at baseline in both smokeless tobacco users and smokers, indicating stereoselective retention of (S)-NNAL. Collectively, the results of this study suggest that there is a receptor in the human body, possibly in the lung, for (S)-NNAL, the more carcinogenic NNAL enantiomer. These data may have considerable implications for understanding mechanisms of tumor induction by NNK.