EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
The cytotoxic effect of purine riboside on COS-7 cells.
PMID 11783515
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I100
5-Iodotubericidin, ≥90%, solid
C11H13IN4O4