EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Synthesis of taurine analogues from alkenes: preparation of 2-aminocycloalkanesulfonic acids.
PMID 11787625
Abstract
(+/-)trans 2-Aminocyclohexanesulfonic acid and (+/-)trans 2-aminocyclopentanesulfonic acid were prepared from cyclohexene and cyclopentene respectively by sulfur monochloride addition, followed by oxidation to 2-chlorosulfonic acid and substitution of chlorine.