EMAIL THIS PAGE TO A FRIEND
Biomaterials
Characterization of porous collagen/hyaluronic acid scaffold modified by 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide cross-linking.
PMID 11791924
Abstract
In order to develop a scaffolding material for tissue regeneration, porous matrices containing collagen and hyaluronic acid were fabricated by freeze drying at -20 degrees C, -70 degrees C or -196 degrees C. The fabricated porous membranes were cross-linked using 1-ethyl-3-(3-dimethyl aminopropyl) carbodiimide (EDC) in a range of 1-100 mM concentrations for enhancing mechanical stability of the composite matrix. Scanning electron microscope (SEM) views of the matrices demonstrated that the matrices obtained before cross-linking process had interconnected pores with mean diameters of 40, 90 or 230 microm and porosity of 58-66% according to the freezing temperature, and also the porous structures after cross-linking process were retained. The swelling test and IR spectroscopic measurement of different cross-linked membranes were carried out as a measure of the extent of cross-linking. The swelling behavior of cross-linked membranes showed no significant differences as cross-linking degree increased. FT-IR spectra showed the increase of the intensity of the absorbencies at amide bonds (1655, 1546, 1458 cm(-1)) compared to that of CH bond (2930 cm(-1)). In enzymatic degradation test, EDC treated membranes showed significant enhancement of the resistance to collagenase activity in comparison with 0.625% glutaraldehyde treated membranes. In cytotoxicity test using L929 fibroblastic cells, the EDC-cross-linked membranes demonstrated no significant toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
165344
1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide methiodide
C9H20IN3