EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Carnitine biosynthesis in mammals.
PMID 11802770
Abstract
Carnitine is indispensable for energy metabolism, since it enables activated fatty acids to enter the mitochondria, where they are broken down via beta-oxidation. Carnitine is probably present in all animal species, and in numerous micro-organisms and plants. In mammals, carnitine homoeostasis is maintained by endogenous synthesis, absorption from dietary sources and efficient tubular reabsorption by the kidney. This review aims to cover the current knowledge of the enzymological, molecular, metabolic and regulatory aspects of mammalian carnitine biosynthesis, with an emphasis on the human and rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
403245
(3-Carboxypropyl)trimethylammonium chloride, technical grade
C7H16ClNO2