EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
Detection of beta-ureidopropionase deficiency with HPLC-electrospray tandem mass spectrometry and confirmation of the defect at the enzyme level.
PMID 11804209
Abstract
The pyrimidine bases uracil and thymine are degraded via the consecutive action of three enzymes to beta-alanine and beta-aminoisobutyric acid, respectively. To date, a number of patients have been described with a deficiency of dihydropyrimidine dehydrogenase and dihydropyrimidinase, the first two enzymes of the pyrimidine degradation pathway. In this study, we demonstrate that the first patient presenting with N-carbamyl-beta-amino aciduria, due to a deficiency of beta-ureidopropionase, was easily diagnosed at the metabolite level using HPLC-tandem mass spectrometry. Urinary analysis showed strongly elevated levels of N-carbamyl-beta-alanine and N-carbamyl-beta-aminoisobutyric acid, with normal or moderately increased levels of the pyrimidine bases and the dihydropyrimidines, respectively. The deficiency of beta-ureidopropionase was confirmed by measuring all three enzymes of the pyrimidine degradation pathway. No activity of beta-ureidopropionase could be detected in a liver biopsy of the patient, while a normal activity of dihydropyrimidine dehydrogenase and dihydropyrimidinase was present. Thus, HPLC-tandem mass specrometry proved to be a powerful tool for the initial diagnosis of patients with deficiency of beta-ureidopropionase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217794
DL-3-Aminoisobutyric acid, 98%
C4H9NO2