EMAIL THIS PAGE TO A FRIEND
Applied microbiology
Isolation of acetyl T-2 toxin from Fusarium poae.
PMID 1180555
Abstract
Acetyl T-2 toxin (3,4,15-triacetoxy-8-isovaleroxy-12,13-epoxy-delta9-trichothecene) was isolated and characterized as a naturally occurring emetic trichothecene from liquid cultures of Fusarium poae (NRRL 3287). Acetyl T-2 toxin was shown to be much less toxic than T-2 toxin in pigeon assays.