EMAIL THIS PAGE TO A FRIEND
Animal biotechnology
Comparative mapping of RPL3, a gene overexpressed in multiple obesity models.
PMID 11808632
Abstract
The ribosomal protein 3 gene is differentially expressed in hypothalamus and brown adipose tissue between mouse lines divergently selected for heat loss, and in skeletal muscle of the ob/ob mouse model. Unfortunately, multiple Rpl3-processed pseudogenes have hampered mapping of the functional gene copy in mammalian species. Using PCR amplification with intronic primer binding, we have mapped Rpl3 to MMU15, and have also localized RPL3 to BTA5 in cattle. Comparative mapping implicates a previously mapped copy of RPL3 on HSA22 as the functional copy of human RPL3, while predictive mapping places the porcine homologue on SSC5.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV48157
Anti-RPL3 antibody produced in rabbit, affinity isolated antibody