EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cryphonectric acid and other minor metabolites from a hypovirulent strain of Cryphonectria parasitica.
PMID 11809063
Abstract
Investigations carried out on secondary metabolites produced in culture by a hypovirulent strain of Cryphonectria parasitica allowed the isolation of several compounds which were characterized by NMR analysis and derivatization reactions. The most abundant metabolite was a new compound, called cryphonectric acid (1). Other metabolites were diaporthin, the only known phytotoxic compound isolated from both virulent and hypovirulent strains of C. parasitica, (+)-orthosporin, and L-p-hydroxyphenyllactic acid (HOPLA). Root growth activity of the purified compounds was evaluated both on tomato seedlings and maize subapical segments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3253
DL-p-Hydroxyphenyllactic acid, ≥97% (HPLC)
C9H10O4