EMAIL THIS PAGE TO A FRIEND
Histochemistry and Cell Biology
Toluidine blue-O staining of prion protein deposits.
PMID 11810193
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5058
Anti-Prion Protein Antibody, NT, a.a. 78-97, serum, clone, Chemicon®