EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Solid-phase synthesis of alpha-substituted 3-bisarylthio N-hydroxy propionamides as specific MMP inhibitors.
PMID 11814839
Abstract
A novel series of potent and specific MMP-2,3,9,13 inhibitors has been obtained by modulation on solid phase by alpha and aryl substitutions on 3-arylthio-N-hydroxy-propionamides starting from itaconic acid.
Related Materials