EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Long-acting fentanyl analogues: synthesis and pharmacology of N-(1-phenylpyrazolyl)-N-(1-phenylalkyl-4-piperidyl)propanamides.
PMID 11814871
Abstract
The synthesis of new fentanyl analogues in which the benzene ring of the propioanilido group has been replaced by phenylpyrazole is described. Antinociceptive activity was evaluated using the writhing and hot plate tests in mice. Two compounds, and, showed interesting analgesic properties, being more potent than morphine and less than fentanyl but with longer duration of action. These compounds inhibited the electrically evoked muscle contraction of guinea pig ileum and mouse vas deferens but not that of rabbit vas deferens and the effects could be reversed by antagonists (naloxone and/or CTOP), thus indicating that the compounds acted as mu agonists. Finally, the binding data confirmed that the compounds had high affinity and selectivity for the mu receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131385
1-Phenethyl-4-piperidone, 98%
C13H17NO