EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Thrombostatin inhibits cyclic flow variations in stenosed canine coronary arteries.
PMID 11816721
Abstract
Thrombostatins are a group of compounds based upon a breakdown product of bradykinin, RPPGF. They inhibit alpha-thrombin-induced platelet activation by binding to protease activated receptor 1 and, at a lower affinity, by interacting with thrombin's active site. After a single intravenous infusion of MAP4-RPPGF (11.58 mg/kg), its t1/2alpha was 4.5 min with a clearance of 2.0 ml/min. MAP4-RPPGF administration had a sustained antiplatelet effect, preventing gamma-thrombin-induced (12.5 nM) platelet activation for 4 h. Its antiplatelet effect summated with that of aspirin and/or clopidogrel. MAP4-RPPGF was compared with aspirin and clopidogrel in the Folts model of coronary artery thrombosis. Dogs were randomized to 3 treatment groups: aspirin 1.14 mg/kg i.v., clopidogrel 0.5 mg/kg i.v., or MAP4-RPPGF 0.77 mg/kg i.v. Cyclic flow variations (CFV) were recorded in 5 untreated dogs hourly for 3 successive hours and for 1 h before (all groups >11 CFV/h), and for 2 h after drug infusion in each of the 3 treatment groups. After 1 h drug treatment, all groups of animals had <6 CFV/h; after 2 h treatment, all had <1 CFV/h. All agents significantly reduced CFV from control at each hour, but none was significantly better than any other. Thrombostatin was as effective as aspirin or clopidogrel in inhibiting coronary artery thrombosis in this canine model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1401
Bradykinin Fragment 1-5, ≥97% (HPLC)
C27H40N8O6