EMAIL THIS PAGE TO A FRIEND
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan
[Effects of organonitrogen, carbamate pesticides and others on beta-hexosaminidase release from rat basophilic leukemia cells (RBL-2H3)].
PMID 11817142
Abstract
There have been very few reports of the effect of pesticides on immediate allergy. In the previous report, the effect of pyrethroid pesticides and organophosphorus pesticides on immediate allergic reaction was investigated. Subsequently, 12 organonitrogen pesticides, 14 carbamate pesticides and 4 other pesticides were investigated for their effects on the enzyme activity of beta-hexosaminidase as an index of chemical mediator release from rat basophilic leukemia cells (RBL-2H3). Two organonitrogen pesticides, bitertanol and pyridaben, and two organotin pesticides, cyhexatin and fenbutatin oxide, were found to promote beta-hexosaminidase release. Bitertanol non-specifically caused the promotion of chemical mediator release, while the release-promotive action of pyridaben was related to IgE antibody and those of cyhexatin and fenbutatin oxide to cell injury. On the other hand, two organonitrogen pesticides, propiconazole and triadimenol, and imazalil showed release-inhibitory action. These data suggested that some pesticides can affect immediate allergy.
Related Materials