EMAIL THIS PAGE TO A FRIEND
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan
[Method-performance studies of notified analytical method for chinomethionat].
PMID 11817145
Abstract
Method-performance studies were conducted for the notified revised analytical method of chinomethionat in agricultural products by interlaboratory study. Six laboratories analyzed unpolished rice, cabbage, squash, lettuce and orange spiked with 0.1 microgram/g of chinomethionat in replicate. Mean values of recovery from the 5 crops ranged from 90.2 to 100.5%. Repeatability relative standard deviations ranged from 4.4 to 7.7% and reproducibility relative standard deviations ranged from 10.9 to 17.1%. The detection limits were 0.003-0.012 microgram/g.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45372
Chinomethionate, PESTANAL®, analytical standard
C10H6N2OS2