EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Pharmacology of tetrandrine and its therapeutic use in digestive diseases.
PMID 11819843
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001165
Tetrandrine, European Pharmacopoeia (EP) Reference Standard
C38H42N2O6