EMAIL THIS PAGE TO A FRIEND
Organic letters
Unexpected remarkable stability of primary ozonides derived from alkenyl stannanes. One-pot synthesis of 1,2-diols from alkynes.
PMID 11820885
Abstract
Primary ozonides derived from alkenyl stannanes display an unusual stability and can be transformed into 1,2-diols by treatment with dimethyl sulfide and borane-methyl sulfide complex. This observation has been incorporated into the development of a novel one-pot strategy for the conversion of alkynes into 1,2-diols.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
192120
Borane dimethyl sulfide complex solution, 2.0 M in THF
C2H9BS