EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Anticonvulsant activity of reaction products of 5,5-diphenylhydantoin with substituted methylene bromides.
PMID 11822175
Abstract
Some derivatives of 5,5-diphenylhydantoin (phenytoin) were synthesized by the alkylation of phenytoin with substituted methylene bromides. The hydantoins were evaluated for possible anticonvulsant activity in the maximal electroshock (MES)- and subcutaneous pentylenetetrazole (ScMet)-induced seizures and for neurotoxicity in the rotorod test in mice and rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D41686
Dibromomethane, 99%
CH2Br2
47799
Dibromomethane, analytical standard
CH2Br2