EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Determination of triacontanol and octacosanol in fenglasu and fenglasu capsules by capillary gas chromatography].
PMID 11822291
Abstract
The method was established for the determination of triacontanol and octacosanol in fenglasu and fenglasu capsules. The solvent was chloroform. The internal standard was hexatriacontanol. Using SIMPLCITY-1 capillary column, flame ionization detector was specified, injector temperature was 300 degrees C, and the column temperature rise by programming. [Initial temperature was 240 degrees C, maintaining 1 min and then rising the temperature (5 degrees C.min-1) to 300 degrees C for 15 min]. The flow rate of Nitrogen was 27 ml.min-1. The calibration curves were linear in the range of 0.4-1.6 mg/ml for triacontanol(r = 0.999), 0.2-0.6 mg/ml for octacosanol(r = 0.9996) respectively. The average recovery (n = 9) was 99.2% for triacontanol (RSD = 1.15%), 99.91% for octacosanol (RSD = 1.40%). The method is rapid, accurate and reliable for the quality control of fenglasu and fenglasu capsules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3379
1-Octacosanol, ≥99% (GC)
C28H58O
T3777
1-Triacontanol, ≥98% (capillary GC)
C30H62O