EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Tuning the activity of catechol oxidase model complexes by geometric changes of the dicopper core.
PMID 11822456
Abstract
Dicopper(II) complexes of a series of different pyrazolate-based dinucleating ligands [L1](-)-[L4](-) have been synthesized and characterized structurally and spectroscopically. A major difference between the four complexes is the individual metal-metal separation that is enforced by the chelating side arms of the pyrazolate ligand scaffold: it varies from 3.45 A in 2 x (BF4)4 to 4.53 A in 4 x (ClO4)2. All complexes have been evaluated as model systems for the catechol oxidase enzyme by using 3,5-di-tert-butylcatechole (DTBC) as the test substrate. They were shown to exhibit very different catecholase activities ranging from very efficient to poor catalysts (k(obs) between 2430+/-202 and 22.8+/-1.2 h(-1)), with an order of decreasing activity 2 x (ClO4)4 > 1 x (ClO4)2 > 3 x (ClO4)2 > 4 x (ClO4)2. A correlation of the catecholase activities with the variation in Cu...Cu distances, as well as other effects resulting from the distinct redox potentials, neighboring groups, and the individual coordination spheres are discussed. Saturation behavior for the rate dependence on substrate concentration was observed in only two cases, that is, for the most active 2 x (ClO4)4 and for the least active 4 x (ClO4)2, whereas a catalytic rate that is almost independent of substrate concentration (within the range studied) was observed for 1 x (ClO4)2 and 3 x (ClO4)2. H2O2 was detected as the product of O2 reduction in the catecholase reaction of the three most active systems. The structures of the adducts of "L3Cu2" and "L4Cu2" with a substrate analogue (tetrachlorocatecholate, TCC) suggest a bidentate substrate coordination to only one of the copper ions for those catalysts that feature short ligand side arms and correspondingly exhibit larger metal-metal separations; this possibly contributes to the lower activity of these systems. TCC binding is supported by several H-bonding interactions to water molecules at the adjacent copper or to ligand-side-arm N-donors; this emphasizes the importance of functional groups in proximity to the bimetallic active site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D45800
3,5-Di-tert-butylcatechol, 98%
C14H22O2