EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Induction of glutathione synthesis by oxidized low-density lipoprotein and 1-palmitoyl-2-arachidonyl phosphatidylcholine: protection against quinone-mediated oxidative stress.
PMID 11829739
Abstract
Exposure of endothelial cells to oxidized low-density lipoprotein (oxLDL) leads to diverse cellular effects, including induction of the intracellular antioxidant GSH. It is not known whether lipid-or protein-derived oxidation products cause GSH induction and whether this involves increased activity of the key enzyme in its synthesis, glutamate-cysteine ligase (GCL). Furthermore, the effect of oxLDL exposure on the cell's ability to combat oxidative stress has not been previously examined. In the present study we found that, in bovine aortic endothelial cells, LDL or 1-palmitoyl-2-arachidonyl phosphatidylcholine oxidized by different reactive oxygen and nitrogen species induced GSH synthesis. However, prevention of GSH synthesis during exposure to oxLDL caused extensive cell death. The mediator causing GSH induction was shown to be a polar lipid and resulted in the increased activity of GCL as well as increased protein levels of the regulatory subunit of GCL. Pretreatment with both oxLDL and the polar lipid subfraction of the oxLDL protected cells against the toxicity of 2,3-dimethoxynaphthoquinone (DMNQ), a superoxide- and H(2)O(2)-forming compound. The potential of a low level of lipid peroxidation products to initiate cytoprotective pathways are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0319
2-Arachidonoyl-1-palmitoyl-sn-glycero-3-phosphocholine, 10 mg/mL in chloroform
C44H80NO8P