EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
In vitro effect of bioactive compounds on influenza virus specific B- and T-cell responses.
PMID 11841689
Abstract
In vitro studies have demonstrated positive effects of bioactive compounds on several functions of the immune system. In the present study, 25 of such compounds were tested for their immune modulating properties on influenza virus specific human B- and T-cell responses in vitro. One of these compounds, N-acetyl-L-cysteine was shown to increase influenza virus specific lymphocyte proliferation and interferon(IFN)-gamma production at a concentration of 1.0 mmol/l. Furthermore, N-acetyl-L-cysteine was found to enhance a specific activity of two influenza specific CD8+ cytotoxic T-lymphocyte clones directed towards HLA-A*0201 and HLA-B*2705 restricted epitopes. A second compound, chlorogenic acid, was shown to enhance antigen specific proliferation of lymphocytes in three out of four donors, at concentrations of 10-50 micromol/l. Neither of the two compounds exhibited a positive effect on the production of influenza virus specific antibodies by human peripheral blood mononuclear cells in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3251
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
C29H50O2
C9538
Coenzyme Q10, ≥98% (HPLC)
C59H90O4