EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
New methods for the synthesis of N-benzoylated uridine and thymidine derivatives; a convenient method for N-debenzoylation.
PMID 11841817
Abstract
An improved procedure for the synthesis of N-benzoyl-2',3'-O-isopropylidene uridine via one-step selective N-benzoylation of 2',3' -O-isopropylidene uridine has been developed. An efficient synthetic route to N-benzoyl thymidine via initial tribenzoylation, followed by selective hydrolysis of the benzoates is also described. De-N-benzoylation of N-benzoylated thymidine and uridine derivatives can be conveniently effected under neutral conditions, by heating with benzyl alcohol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5127
2′,3′-O-Isopropylideneuridine, ≥99% (HPLC)
C12H16N2O6