EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Orexin receptor-1 (OX-R1) immunoreactivity in chemically identified neurons of the hypothalamus: focus on orexin targets involved in control of food and water intake.
PMID 11849298