EMAIL THIS PAGE TO A FRIEND
Current opinion in biotechnology
Biocatalytic synthesis of hydroxylated natural products using aldolases and related enzymes.
PMID 11849940
Abstract
Synthetic building blocks bearing hydroxylated chiral centers are important targets for biocatalysis. Many C-C bond forming enzymes have recently been investigated for new applications and new strategies towards the synthesis of natural products and related oxygenated compounds. Several old catalysts have been studied to increase our functional knowledge of natural aldolase-type enzymes, and new mutated catalysts or catalytic antibodies have been tested for their synthetic utility.
Related Materials