EMAIL THIS PAGE TO A FRIEND
The Journal of Investigative Dermatology
Matrix metalloproteinase inhibitor BB-3103 unlike the serine proteinase inhibitor aprotinin abrogates epidermal healing of human skin wounds ex vivo.
PMID 11851876