EMAIL THIS PAGE TO A FRIEND
Clinical nurse specialist CNS
Controlled-release oxycodone hydrochloride (OxyContin).
PMID 11855610
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O1378
Oxycodone hydrochloride, analytical standard
C18H21NO4 · HCl