EMAIL THIS PAGE TO A FRIEND
Transplantation
Induction of rejection of successful allografts of rat islets by donor peritoneal exudate cells.
PMID 118559
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
118559
1,5-Hexadien-3-ol
C6H10O