EMAIL THIS PAGE TO A FRIEND
Hong Kong medical journal = Xianggang yi xue za zhi
Topical chloramphenicol for eye infections.
PMID 11861993
Abstract
Topical chloramphenicol has been widely used in the treatment and prevention of superficial eye infections due to its broad spectrum of activity and low cost. The use of this drug has decreased considerably in the United States since the first case of aplastic anaemia associated with topical chloramphenicol was reported in the 1960s. This medication, however, is still widely used in many other countries. This paper evaluates the evidence for and against the use of topical chloramphenicol in ocular diseases.