EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Molecular basis for the inhibition of hypoxia-induced apoptosis by 2-deoxy-D-ribose.
PMID 11866437
Abstract
An angiogenic factor, platelet-derived endothelial cell growth factor/thymidine phosphorylase (PD-ECGF/TP), stimulates the chemotaxis of endothelial cells and confers resistance to apoptosis induced by hypoxia. 2-deoxy-D-ribose, a degradation product of thymidine generated by TP enzymatic activity partially prevented hypoxia-induced apoptosis. 2-Deoxy-D-ribose inhibits a number of components of the caspase-mediated hypoxia-induced apoptotic pathway. It inhibits hypoxia-induced caspase 3 activation, mitochondrial cytochrome c release, downregulation of Bcl-2 and Bcl-x(L), upregulation of hypoxia-inducible factor (HIF)-1 alpha, and loss of mitochondrial transmembrane potential in human leukemia HL-60 cell line. These findings suggest a molecular mechanism by which 2-deoxy-d-ribose confers the resistance to apoptosis. Thus 2-deoxy-D-ribose-modulated suppression of HIF-1 alpha expression could prevent the hypoxia-induced decrease of the anti-apoptotic Bcl-2 and Bcl-x(L) on the mitochondria. 2-Deoxy-L-ribose and its analogs may enhance apoptosis and suppress the growth of tumors by competitively inhibiting the activities of 2-deoxy-d-ribose and thus these analogs show promise for anti-tumor therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75617
2-Deoxy-L-ribose, ≥97.0% (TLC)
C5H10O4