EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Diazo group electrophilicity in kinamycins and lomaiviticin A: potential insights into the molecular mechanism of antibacterial and antitumor activity.
PMID 11866589
Abstract
Theoretical and chemical studies of the reactivity of isoprekinamycin, the kinamycins, and the lomaiviticins support the proposal that these natural products exhibit enhanced diazonium salt character and may owe their antitumor antibiotic properties to their ability to act as electrophilic azo-coupling agents in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12490
11H-Benzo[a]fluorene, ≥98.0%
C17H12