EMAIL THIS PAGE TO A FRIEND
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury
[Balneological use of bath concentrate containing xidifon and essential oils from ginger, nutmeg, and sandal].
PMID 11868534
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W300500 Sandalwood oil, natural