EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Iminodibenzyl as a novel coupling agent for the spectrophotometric determination of sulfonamide derivatives.
PMID 11880002
Abstract
A rapid, selective and simple spectrophotometric method for the determination of sulfa-drugs is described. The method is based on the formation of violet colored azo product by the diazotization of sulfonamides, viz. sulfathiazole (SFT), sulfadiazine (SFD), sulfacetamide (SFA), sulfamethoxazole (SFMx), sulfamerazine (SFMr), sulfaguanidine (SFG) and sulfadimidine (SFDd) followed by a coupling reaction with iminodibenzyl in alcohol medium. Absorbance of the resulting violet azo product is measured at 570-580 nm and is stable for 24 h at 27 degrees C. Beer's law is obeyed in the concentration range of 0.05-6.0 microg ml(-1) at the wavelength of maximum absorption. The method is successfully employed for the determination of sulfonamides in various pharmaceutical preparations and common excipients used as additives in pharmaceuticals do not interfere in the proposed method. The method offers the advantages of simplicity, rapidity and sensitivity without the need for extraction or heating. A reaction mechanism is proposed for the formation of the violet azo product.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D34108
Dibenzylamine, 97%
C14H15N