EMAIL THIS PAGE TO A FRIEND
Acta bio-medica de L'Ateneo parmense : organo della Societa di medicina e scienze naturali di Parma
[Specific inhibitors of cyclooxygenase-2 (COX-2): current knowledge and perspectives].
PMID 11889908
Abstract
The Authors summarize the current knowledge on a new class of nonsteroidal anti-inflammatory drugs (NSAIDs), the coxib (celecoxib and rofecoxib), in the treatment of rheumatic diseases. Celecoxib and rofecoxib are selective cyclooxygenase-2 (COX-2) inhibitors which possess the same anti-inflammatory and analgesic activities, but a better gastric tolerability compared to the non-selective COX-1 and COX-2 inhibitors. The Authors also report other possible therapeutic effects of these NSADIs as evidenced by the more recent data of the literature. Celecoxib seems to reduce the incidence of new polyps in patients with familial adenomatous polyposis. It has been suggested the use of celecoxib as a protective drug against the development of colorectal cancer. Other (neoplastic) or pre-neoplastic conditions, such as bladder dysplasia, Barret esophagus, attinic keratosis and Alzheimer's disease seem to have benefit from this class of drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7742
Prostaglandin G2, ≥95% (HPLC), acetone solution
C20H32O6