EMAIL THIS PAGE TO A FRIEND
Zhurnal evoliutsionnoi biokhimii i fiziologii
[Ammonium compounds with localized and nonlocalized charges as reversible inhibitors of choline esterases of different origin].
PMID 11898593
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2880
Propionylthiocholine iodide, ≥98%
C8H18INOS